Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtpbp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159329.1 | NP_001152801.1 | 2 | 200 | ||
NM_025332.3 | NP_079608.1 | 2 | 401 | |||
GenBank mRNA | AK002771.1 | - | 2 | 386 | ||
AK010979.1 | - | 2 | 398 | |||
AK078242.1 | - | 1 | 907 | |||
AK147840.1 | - | 2 | 385 | |||
BC107216.1 | - | 2 | 398 | 40051675 | ||
BC107217.1 | - | 2 | 197 | 40051680 |