Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snupn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178374.3 | NP_848461.1 | 9 | 816 | ||
GenBank mRNA | AK002717.1 | - | 9 | 832 | ||
AK155315.1 | - | 9 | 833 | |||
BC049638.1 | - | 9 | 816 | 6589249 | ||
BC061258.1 | - | 9 | 821 | 30251661 |