Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ethe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023154.3 | NP_075643.1 | 2, 3 | 654 | ||
GenBank mRNA | AB049623.1 | - | 1, 2 | 107 | ||
AK002666.1 | - | 1, 2 | 109 | |||
BC010592.1 | - | 1, 2 | 112 | 3156338 | ||
BC083162.1 | - | 1, 2 | 88 | 6529658 | ||
BC094044.1 | - | 2, 3 | 654 | 5064931 |