Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000427.2 | NP_000418.2 | 2 | 1098 | ||
GenBank mRNA | BC034690.1 | - | 1 | 1089 | 4752776 | |
BC108290.1 | - | 2 | 1110 | 4754528 | ||
M61120.1 | - | 1 | 1089 |