Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164823.1 | NP_001158295.1 | 2 | 337 | ||
NM_025339.4 | NP_079615.1 | 2 | 337 | |||
GenBank mRNA | AK002688.1 | - | 2 | 313 | ||
AK012820.1 | - | 2 | 312 | |||
BC049583.1 | - | 2 | 337 | 6742203 |