Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rmnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025343.5 | NP_079619.4 | 5 | 868 | ||
GenBank mRNA | AK007323.1 | - | 5 | 816 | ||
AK075603.1 | - | 5 | 851 | |||
AK163523.1 | - | 104 | ||||
BC019572.1 | - | 2 | 113 | |||
BC027299.1 | - | 5 | 816 | 3987394 | ||
BC036150.1 | - | 4 | 788 | 4977394 |