Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rmnd5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025346.1 | NP_079622.1 | 8 | 1262 | ||
GenBank mRNA | AK005344.1 | - | 5 | 641 | ||
AK075599.1 | - | 8 | 1262 | |||
AK179668.1 | - | 2 | 124 | |||
BC016075.1 | - | 8 | 1116 | 4485143 |