Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ypel3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025347.2 | NP_079623.1 | 5 | 887 | ||
NM_026875.2 | NP_081151.2 | 5 | 987 | |||
GenBank mRNA | AF523353.1 | - | 5 | 877 | ||
AK002925.1 | - | 5 | 871 | |||
AK003371.1 | - | 5 | 888 | |||
AK004431.1 | - | 5 | 960 | |||
BC009171.1 | - | 5 | 861 |