Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025356.4 | NP_079632.1 | 5 | 401 | ||
GenBank mRNA | AK003163.1 | - | 5 | 352 | ||
AK004001.1 | - | 5 | 411 | |||
AK009276.1 | - | 5 | 343 | |||
AK009936.1 | - | 5 | 340 | |||
AK032885.1 | - | 5 | 411 | |||
AK049906.1 | - | 5 | 402 | |||
AK077666.1 | - | 5 | 461 | |||
AK088448.1 | - | 5 | 371 | |||
AK151295.1 | - | 5 | 515 | |||
AK151594.1 | - | 5 | 422 | |||
AK152231.1 | - | 5 | 422 | |||
AK152541.1 | - | 5 | 395 | |||
AK153100.1 | - | 5 | 515 | |||
AK153111.1 | - | 5 | 423 | |||
AK153410.1 | - | 5 | 422 | |||
AK153423.1 | - | 5 | 461 | |||
AK159125.1 | - | 5 | 461 | |||
AK159546.1 | - | 5 | 397 | |||
AK159963.1 | - | 5 | 395 | |||
AK165235.1 | - | 4 | 642 | |||
AK167077.1 | - | 5 | 411 | |||
AK167138.1 | - | 5 | 364 | |||
AK167650.1 | - | 5 | 399 | |||
AK167801.1 | - | 5 | 423 | |||
AK169177.1 | - | 4 | 773 | |||
AK208479.1 | - | 4 | 182 | |||
BC023266.1 | - | 4 | 166 | |||
BC024351.1 | - | 6 | 450 | |||
BC057941.1 | - | 5 | 382 | 5323636 | ||
BC098220.1 | - | 5 | 416 | 963757 |