Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025356.4 | NP_079632.1 | 7 | 609 | ||
GenBank mRNA | AK003163.1 | - | 7 | 560 | ||
AK004001.1 | - | 7 | 619 | |||
AK009276.1 | - | 7 | 551 | |||
AK009936.1 | - | 7 | 548 | |||
AK014344.1 | - | 1 | 90 | |||
AK032885.1 | - | 7 | 619 | |||
AK049906.1 | - | 7 | 610 | |||
AK077666.1 | - | 7 | 669 | |||
AK088448.1 | - | 7 | 579 | |||
AK151295.1 | - | 7 | 723 | |||
AK151594.1 | - | 7 | 630 | |||
AK152541.1 | - | 7 | 603 | |||
AK153100.1 | - | 7 | 723 | |||
AK153111.1 | - | 7 | 631 | |||
AK153423.1 | - | 7 | 669 | |||
AK159125.1 | - | 7 | 669 | |||
AK159546.1 | - | 7 | 605 | |||
AK159963.1 | - | 7 | 603 | |||
AK165235.1 | - | 6 | 850 | |||
AK167077.1 | - | 7 | 619 | |||
AK167138.1 | - | 7 | 572 | |||
AK167650.1 | - | 7 | 607 | |||
AK167801.1 | - | 7 | 631 | |||
AK169177.1 | - | 6 | 981 | |||
BC023266.1 | - | 6 | 374 | |||
BC024351.1 | - | 8 | 658 | |||
BC057941.1 | - | 7 | 590 | 5323636 | ||
BC098220.1 | - | 7 | 624 | 963757 |