Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cml1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023160.2 | NP_075649.2 | 2 | 819 | ||
GenBank mRNA | AF163314.1 | - | 2 | 817 | ||
AK002975.1 | - | 1 | 173 | |||
AK003301.1 | - | 2 | 769 | |||
BC010796.1 | - | 2 | 773 | 4223199 |