Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Josd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205070.1 | NP_001191999.1 | 5 | 649 | ||
NM_001205071.1 | NP_001192000.1 | 4 | 537 | |||
NM_001205072.1 | NP_001192001.1 | 4 | 506 | |||
NM_001205073.1 | NP_001192002.1 | 4 | 503 | |||
NM_025368.4 | NP_079644.1 | 4 | 534 | |||
GenBank mRNA | AK002872.1 | - | 4 | 375 | ||
AK003525.1 | - | 4 | 459 | |||
AK186296.1 | - | 1 | 103 | |||
BC018244.1 | - | 5 | 732 | 4224952 | ||
CA326008.1 | - | 4 | 448 |