Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aaed1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025370.2 | NP_079646.1 | 4 | 368 | ||
GenBank mRNA | AK003782.1 | - | 4 | 369 | ||
AK006589.1 | - | 4 | 223 | |||
AK145073.1 | - | 366 | ||||
AK145867.1 | - | 369 | ||||
AK148379.1 | - | 369 | ||||
AK168581.1 | - | 3 | 180 | |||
AK168701.1 | - | 247 |