Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1110008L16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025373.1 | NP_079649.1 | 6 | 2738 | ||
GenBank mRNA | AK003589.1 | - | 6 | 668 | ||
AK008749.1 | - | 2 | 164 | |||
AK031144.1 | - | 6 | 2116 | |||
AK045434.1 | - | 6 | 2106 | |||
AK151976.1 | - | 6 | 2738 | |||
AK152028.1 | - | 6 | 2738 | |||
AK172942.1 | - | 6 | 542 | |||
BC034876.1 | - | 6 | 1590 | 1512335 | ||
BC037485.1 | - | 6 | 1094 |