Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wbscr22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025375.3 | NP_079651.2 | 3, 4 | 348 | ||
GenBank mRNA | AF412035.1 | - | 2, 3 | 120 | ||
AK003386.1 | - | 2, 3 | 121 | |||
AK011005.1 | - | 2, 3 | 128 | |||
AK151986.1 | - | 2, 3 | 129 | |||
AK178657.1 | - | 1 | 15 | |||
BC093484.1 | - | 2, 3 | 98 | 6735279 | ||
BC117986.1 | - | 3, 4 | 348 | 40102067 | ||
BC117987.1 | - | 3, 4 | 348 | 40102068 |