Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ska2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025377.3 | NP_079653.1 | 3 | 181 | ||
GenBank mRNA | AK003196.1 | - | 3 | 209 | ||
AK013317.1 | - | 3 | 182 | |||
AK013370.1 | - | 3 | 182 | |||
AK017811.1 | - | 3 | 182 | |||
AK153219.1 | - | 152 | ||||
AK162628.1 | - | 178 | ||||
BC052151.1 | - | 182 | ||||
BC060088.1 | - | 184 | ||||
BC096627.1 | - | 193 | ||||
BC117001.1 | - | 3 | 211 | 40126320 | ||
BC119167.1 | - | 3 | 211 | 8733916 |