Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006151.2 | NP_006142.1 | 4 | 597 | ||
NR_027647.1 | 3 | 509 | ||||
XM_011524808.2 | XP_011523110.1 | 4 | 473 | |||
GenBank mRNA | AK131496.1 | - | 3 | 1260 | ||
AK303903.1 | - | 3 | 344 | |||
AK308378.1 | - | 4 | 322 | |||
AK310965.1 | - | 4 | 352 | |||
BC107166.1 | - | 4 | 468 | 40034365 | ||
BC107167.1 | - | 4 | 468 | 40034371 | ||
BX648309.1 | - | 4 | 435 | |||
U39573.1 | - | 4 | 362 |