Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025379.2 | NP_079655.1 | 3 | 931 | ||
GenBank mRNA | BC089538.1 | - | 3 | 931 | 6533206 |