Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eef1e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025380.2 | NP_079656.1 | 3, 4 | 419 | ||
GenBank mRNA | AK003335.1 | - | 3, 4 | 402 | ||
AK009889.1 | - | 3, 4 | 391 | |||
BC048466.1 | - | 3, 4 | 401 | 6589105 | ||
EF437326.1 | - | 1, 2 | 22 |