Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Necap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025383.3 | NP_079659.1 | 5, 6 | 531 | ||
GenBank mRNA | AK003463.1 | - | 5, 6 | 498 | ||
AK009714.1 | - | 1, 2 | 76 | |||
AK145874.1 | - | 5, 6 | 519 | |||
AK155871.1 | - | 5, 6 | 519 | |||
AK207758.1 | - | 2, 3 | 134 | |||
AK213306.1 | - | 2, 3 | 134 | |||
BC037069.1 | - | 5, 6 | 530 | 5002765 |