Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prr13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170911.1 | NP_001164382.1 | 4 | 1019 | ||
NM_025385.3 | NP_079661.1 | 5 | 1137 | |||
GenBank mRNA | AK002430.1 | - | 4 | 987 | ||
AK003850.1 | - | 5 | 1134 | |||
AK008611.1 | - | 3 | 960 | |||
AK008894.1 | - | 5 | 1097 | |||
AK154583.1 | - | 5 | 1097 | |||
AK211617.1 | - | 83 | ||||
BC016234.1 | - | 4 | 1284 | 4503179 |