Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160102.1 | NP_001153574.1 | 11 | 2046 | ||
NM_006151.2 | NP_006142.1 | 13 | 2295 | |||
NR_027647.1 | 12 | 2207 | ||||
XM_011524808.2 | XP_011523110.1 | 11 | 1757 | |||
XM_011524810.2 | XP_011523112.1 | 7 | 1770 | |||
GenBank mRNA | AK303903.1 | - | 12 | 2042 | ||
BC107166.1 | - | 13 | 2166 | 40034365 | ||
BC107167.1 | - | 13 | 2166 | 40034371 | ||
BX538344.1 | - | 9 | 1366 | |||
BX648309.1 | - | 13 | 2133 | |||
EF579964.1 | - | 11 | 1768 | |||
M58151.1 | - | 5 | 816 | |||
U39573.1 | - | 13 | 2060 |