Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025386.3 | NP_079662.1 | 1 | 104 | ||
GenBank mRNA | AK003856.1 | - | 1 | 104 | ||
AK008324.1 | - | 1 | 101 | |||
AK036072.1 | - | 1 | 81 | |||
BC049581.1 | - | 1 | 81 | 6772363 | ||
BY708252.1 | - | 1 | 101 |