Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pop4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025390.4 | NP_079666.1 | 7 | 754 | ||
GenBank mRNA | AK002855.1 | - | 7 | 755 | ||
AK003293.2 | - | 7 | 755 | |||
AK003908.1 | - | 7 | 768 | |||
AK054466.1 | - | 787 | ||||
AK150459.1 | - | 7 | 761 | |||
AK152948.1 | - | 7 | 761 | |||
BC011465.1 | - | 7 | 770 | 3966371 |