Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bola2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175103.3 | NP_780312.1 | 2, 3 | 207 | ||
GenBank mRNA | AK002662.1 | - | 2, 3 | 210 | ||
AK003922.1 | - | 2, 3 | 210 | |||
AK028165.1 | - | 2, 3 | 221 | |||
BC045523.1 | - | 2, 3 | 225 | 6397886 |