Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bccip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025392.2 | NP_079668.2 | 6, 7 | 814 | ||
GenBank mRNA | AK003666.1 | - | 6, 7 | 811 | ||
AK010684.1 | - | 6, 7 | 808 | |||
BC048465.1 | - | 6, 7 | 806 | 6534043 |