Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100a14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163525.2 | NP_001156997.1 | 3 | 204 | ||
NM_025393.4 | NP_079669.2 | 3 | 187 | |||
GenBank mRNA | AA726094.1 | - | 3 | 197 | ||
AK003669.1 | - | 3 | 167 | |||
AK019075.1 | - | 3 | 158 | |||
AK132637.1 | - | 2 | 587 | |||
BC025607.1 | - | 2 | 186 | 5151924 |