Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grina | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023168.3 | NP_075657.1 | 7 | 1090 | ||
GenBank mRNA | AF182040.1 | - | 7 | 1073 | ||
AK002347.1 | - | 3 | 266 | |||
AK003920.1 | - | 7 | 1094 | |||
AK196790.1 | - | 1 | 12 | |||
BC010802.1 | - | 7 | 1123 | 4236429 | ||
BC019157.1 | - | 7 | 1111 | |||
BC037667.1 | - | 7 | 1090 | 5376054 |