Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tomm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025394.3 | NP_079670.1 | 3 | 293 | ||
GenBank mRNA | AK003864.1 | - | 3 | 256 | ||
AK008581.1 | - | 3 | 210 | |||
BC055352.1 | - | 3 | 271 | 5011269 | ||
BE647192.1 | - | 2 | 95 | |||
BU555283.1 | - | 3 | 296 | |||
BY773505.1 | - | 3 | 286 |