Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nat9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025400.3 | NP_079676.1 | 4 | 415 | ||
GenBank mRNA | AK003948.1 | - | 4 | 369 | ||
AK009262.1 | - | 3 | 145 | |||
AK020083.1 | - | 4 | 395 | |||
AK082469.1 | - | 4 | 355 | |||
AK148048.1 | - | 4 | 447 | |||
AK154593.1 | - | 4 | 375 | |||
AK186542.1 | - | 4 | 255 | |||
BC011315.1 | - | 4 | 415 | 4013700 |