Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pycrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025412.2 | NP_079688.2 | 4 | 540 | ||
GenBank mRNA | AK002912.1 | - | 4 | 471 | ||
AK013214.1 | - | 4 | 465 | |||
BC026536.1 | - | 4 | 461 | 4987954 |