Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025418.3 | NP_079694.2 | 7 | 788 | ||
GenBank mRNA | AK002611.1 | - | 7 | 787 | ||
AK003213.1 | - | 6 | 705 | |||
AK004323.1 | - | 7 | 860 | |||
AK009227.2 | - | 7 | 777 | |||
AK042917.1 | - | 1 | 23 | |||
BC026752.1 | - | 7 | 781 | 4034998 |