Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013577.1 | NP_001013595.1 | 3 | 253 | ||
GenBank mRNA | AK004256.1 | - | 2 | 126 | ||
AK005245.1 | - | 3 | 235 | |||
AK012023.1 | - | 3 | 173 | |||
AK159116.1 | - | 3 | 241 | |||
AK169537.1 | - | 2 | 155 | |||
BB641174.1 | - | 3 | 253 | |||
BC116204.1 | - | 3 | 148 | 40090152 | ||
BC116205.1 | - | 3 | 148 | 40090155 |