Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif1ax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025437.4 | NP_079713.2 | 5 | 474 | ||
GenBank mRNA | AK005185.1 | - | 5 | 462 | ||
AK007301.1 | - | 5 | 425 | |||
AK012233.1 | - | 5 | 454 | |||
AK012714.1 | - | 5 | 457 | |||
AK021283.1 | - | 5 | 455 | |||
AK030774.1 | - | 478 | ||||
AK031113.1 | - | 486 | ||||
AK077710.1 | - | 5 | 423 | |||
AK090053.1 | - | 440 | ||||
AK134827.1 | - | 399 | ||||
AK134969.1 | - | 399 | ||||
AK165574.1 | - | 440 | ||||
AK166629.1 | - | 5 | 477 | |||
AK166852.1 | - | 5 | 475 | |||
AK207164.1 | - | 4 | 199 | |||
AK217305.1 | - | 4 | 236 | |||
BC027284.1 | - | 5 | 411 | 3156134 |