Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif1ax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025437.4 | NP_079713.2 | 5, 6 | 551 | ||
GenBank mRNA | AK005185.1 | - | 5, 6 | 539 | ||
AK007301.1 | - | 5, 6 | 502 | |||
AK012233.1 | - | 5, 6 | 531 | |||
AK012714.1 | - | 5, 6 | 534 | |||
AK021283.1 | - | 5, 6 | 532 | |||
AK030774.1 | - | 555 | ||||
AK031113.1 | - | 563 | ||||
AK077710.1 | - | 5, 6 | 500 | |||
AK090053.1 | - | 517 | ||||
AK134827.1 | - | 476 | ||||
AK134969.1 | - | 476 | ||||
AK165574.1 | - | 517 | ||||
AK166629.1 | - | 5, 6 | 554 | |||
AK166852.1 | - | 5, 6 | 552 | |||
AK207164.1 | - | 4, 5 | 276 | |||
AK210743.1 | - | 4, 5 | 265 | |||
AK217305.1 | - | 4, 5 | 313 | |||
BC027284.1 | - | 5, 6 | 488 | 3156134 |