Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160145.1 | NP_001153617.1 | 4, 5 | 492 | ||
NM_001160146.1 | NP_001153618.1 | 4, 5 | 388 | |||
NM_025439.3 | NP_079715.1 | 4, 5 | 357 | |||
GenBank mRNA | AK005248.1 | - | 4, 5 | 344 | ||
AK021225.1 | - | 4, 5 | 344 | |||
AK134909.1 | - | 4, 5 | 322 | |||
AK154423.1 | - | 4, 5 | 344 | |||
AK218177.1 | - | 2, 3 | 159 | |||
BC016524.1 | - | 4, 5 | 332 | 3594475 | ||
BC020291.1 | - | 3, 4 | 309 | |||
BC048017.1 | - | 4, 5 | 311 | |||
BQ918002.1 | - | 4, 5 | 579 | |||
CA460035.1 | - | 4, 5 | 394 | |||
CX208138.1 | - | 4, 5 | 388 |