Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nemf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025441.3 | NP_079717.2 | 5 | 451 | ||
GenBank mRNA | AK005212.1 | - | 5 | 425 | ||
AK017826.1 | - | 5 | 441 | |||
AK039497.1 | - | 5 | 418 | |||
AK041458.1 | - | 5 | 425 | |||
AK089000.1 | - | 2 | 92 | |||
AK171652.1 | - | 5 | 425 | |||
BC037106.1 | - | 5 | 424 | |||
BC080716.1 | - | 5 | 444 | |||
BY241581.1 | - | 4 | 325 |