Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osgep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133676.2 | NP_598437.2 | 10 | 1053 | ||
GenBank mRNA | AK005291.1 | - | 10 | 1002 | ||
AK049480.1 | - | 8 | 1541 | |||
AK049955.1 | - | 994 | ||||
AK076246.1 | - | 10 | 1013 | |||
AK088346.1 | - | 10 | 1016 | |||
AK142730.1 | - | 10 | 1053 | |||
AK145181.1 | - | 10 | 981 | |||
AK157554.1 | - | 1004 | ||||
AK166415.1 | - | 10 | 1013 | |||
AK167893.1 | - | 10 | 994 | |||
AK207563.1 | - | 2 | 119 | |||
BC002296.1 | - | 10 | 988 | 3590664 | ||
BC091757.1 | - | 10 | 975 | 4925017 |