Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002336.2 | NP_002327.2 | 15, 16 | 3547 | ||
XM_006719078.3 | XP_006719141.1 | 15, 16 | 3691 | |||
XM_011520671.2 | XP_011518973.1 | 14, 15 | 3297 | |||
XM_017019304.1 | XP_016874793.1 | 13, 14 | 3365 | |||
GenBank mRNA | AB209683.1 | - | 14, 15 | 2999 | ||
AF074264.1 | - | 15, 16 | 3482 | |||
AK074902.1 | - | 5, 6 | 1084 | |||
BC117136.1 | - | 15, 16 | 3439 | 40125687 | ||
BC126405.1 | - | 15, 16 | 3439 | 8992121 | ||
BC143725.1 | - | 15, 16 | 3439 | 9052239 |