Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eapp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025456.3 | NP_079732.1 | 6 | 710 | ||
GenBank mRNA | AK007442.1 | - | 6 | 665 | ||
AK012800.1 | - | 5 | 553 | |||
AK145027.1 | - | 6 | 729 | |||
AK145041.1 | - | 6 | 729 | |||
AY882557.1 | - | 6 | 629 | |||
BC037622.1 | - | 6 | 737 | 5373275 |