Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025461.5 | NP_079737.1 | 2 | 273 | ||
GenBank mRNA | AK007450.1 | - | 2 | 250 | ||
AK007562.1 | - | 2 | 262 | |||
AK007870.1 | - | 2 | 146 | |||
AK008486.1 | - | 2 | 238 | |||
AK040003.1 | - | 2 | 275 | |||
AK054491.1 | - | 2 | 249 | |||
AK082374.1 | - | 2 | 274 | |||
AK087666.1 | - | 274 | ||||
AK088794.1 | - | 274 | ||||
BC005675.1 | - | 237 | 3153356 | |||
BC100533.1 | - | 204 | 30134701 |