Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aamdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177945.1 | NP_001171416.1 | 3 | 328 | ||
NM_001177946.1 | NP_001171417.1 | 3 | 328 | |||
NM_001177947.1 | NP_001171418.1 | 2 | 224 | |||
NM_183251.3 | NP_899074.2 | 2 | 224 | |||
GenBank mRNA | AA065573.1 | - | 2 | 161 | ||
AK007558.1 | - | 2 | 176 | |||
AK008195.1 | - | 2 | 169 | |||
BC026557.1 | - | 3 | 246 | 5027461 | ||
BC062811.1 | - | 3 | 283 | 1264501 | ||
BY259812.1 | - | 3 | 325 |