Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lbhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286103.1 | NP_001273032.1 | 5 | 729 | ||
GenBank mRNA | BI872942.1 | - | 5 | 730 |
Gene Symbol: 1810009A15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025463.3 | NP_079739.1 | 3 | 233 | ||
GenBank mRNA | AK007390.1 | - | 3 | 214 | ||
AK012061.1 | - | 3 | 223 | |||
BC047099.1 | - | 3 | 212 | 5253822 | ||
BC058702.1 | - | 3 | 196 | 6822952 | ||
BY273399.1 | - | 2 | 376 |