Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gkn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025467.1 | NP_079743.1 | 3 | 218 | ||
GenBank mRNA | AK007451.1 | - | 3 | 218 | ||
AK007705.1 | - | 3 | 218 | |||
AK008692.1 | - | 3 | 219 | |||
AK008986.1 | - | 3 | 219 | |||
AY943909.1 | - | 3 | 226 | |||
BC099445.1 | - | 3 | 207 | 30788014 |