Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_078479.3 | NP_510964.1 | 2 | 159 | ||
GenBank mRNA | AB049956.1 | - | 3 | 184 | ||
AK002588.1 | - | 3 | 187 | |||
AK003462.1 | - | 3 | 180 | |||
AK003602.1 | - | 3 | 179 | |||
AK007662.1 | - | 3 | 188 | |||
AK007838.1 | - | 2 | 170 | |||
AK008265.1 | - | 3 | 202 | |||
AK008293.1 | - | 3 | 181 | |||
AK009049.1 | - | 3 | 180 | |||
AK009703.1 | - | 3 | 188 | |||
BC027546.1 | - | 2 | 163 | 1515797 | ||
BC125658.1 | - | 2 | 160 | 40130173 | ||
BC125660.1 | - | 2 | 160 | 40130175 |