Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025473.3 | NP_079749.1 | 8, 9 | 1055 | ||
GenBank mRNA | AK007715.1 | - | 8, 9 | 1056 | ||
AK012353.1 | - | 4, 5 | 383 | |||
BC011237.1 | - | 8, 9 | 1053 | |||
BC019752.1 | - | 9, 10 | 821 | |||
BC054411.1 | - | 8, 9 | 728 | |||
BC069871.1 | - | 8, 9 | 1190 | 6467541 |