Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam53c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175104.4 | NP_780313.1 | 4 | 1295 | ||
GenBank mRNA | AK044469.1 | - | 4 | 1099 | ||
AK135712.1 | - | 3 | 1041 | |||
AK154214.1 | - | 4 | 1099 | |||
AK166841.1 | - | 3 | 1039 | |||
BC044896.1 | - | 3 | 1020 | |||
BC057111.1 | - | 4 | 1295 | 6417613 |