Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam53c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175104.4 | NP_780313.1 | 2, 3 | 477 | ||
GenBank mRNA | AK044469.1 | - | 2, 3 | 281 | ||
AK154214.1 | - | 2, 3 | 281 | |||
BC057111.1 | - | 2, 3 | 477 | 6417613 |