Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpd52l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291197.1 | NP_001278126.1 | 5 | 585 | ||
NM_001291200.1 | NP_001278129.1 | 5 | 585 | |||
NM_001291201.1 | NP_001278130.1 | 4 | 525 | |||
NM_001291202.1 | NP_001278131.1 | 4 | 525 | |||
NM_001291203.1 | NP_001278132.1 | 4 | 525 | |||
NM_001291204.1 | NP_001278133.1 | 3 | 376 | |||
NM_025482.3 | NP_079758.2 | 5 | 585 | |||
GenBank mRNA | AA218395.1 | - | 3 | 294 | ||
AK013085.1 | - | 5 | 527 | |||
AK013188.1 | - | 5 | 538 | |||
AK051268.1 | - | 4 | 469 | |||
AK147885.1 | - | 5 | 545 | |||
AK155987.1 | - | 4 | 462 | |||
AK159925.1 | - | 5 | 513 | |||
AK160727.1 | - | 4 | 469 | |||
AK161092.1 | - | 4 | 481 | |||
AK171825.1 | - | 5 | 547 | |||
BC006886.1 | - | 5 | 474 | 2647702 | ||
CN721218.1 | - | 4 | 439 | |||
DV047279.1 | - | 5 | 571 |