Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025375.2 | NP_001020546.1 | 10 | 999 | ||
NM_001025376.2 | NP_001020547.1 | 10 | 888 | |||
NM_023191.3 | NP_075680.1 | 10 | 893 | |||
GenBank mRNA | AF309554.1 | - | 10 | 851 | ||
AK010283.1 | - | 10 | 839 | |||
AK012342.1 | - | 10 | 905 | |||
AK013114.1 | - | 10 | 857 | |||
AK077037.1 | - | 933 | ||||
AK142498.1 | - | 10 | 856 | |||
AK153862.1 | - | 10 | 926 | |||
AK167706.1 | - | 10 | 926 | |||
AK205840.1 | - | 4 | 285 | |||
AK219807.1 | - | 2 | 54 | |||
BC023026.1 | - | 10 | 838 | 5374890 | ||
EF011623.1 | - | 10 | 836 |